Sodium thiosulfate, 99%, for analysis, anhydrous
Sodium thiosulfate, 99%, for analysis, anhydrous
Sodium thiosulfate, 99%, for analysis, anhydrous
Sodium thiosulfate, 99%, for analysis, anhydrous
Thermo Scientific Chemicals

Sodium thiosulfate, 99%, for analysis, anhydrous

CAS: 7772-98-7 | Na2O3S2 | 158.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 450620050
Price (USD)
180.00
Each
In stock
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)
180.00
Each
Add to cart
Chemical Identifiers
CAS7772-98-7
IUPAC Namedisodium sulfanidesulfonate
Molecular FormulaNa2O3S2
InChI KeyAKHNMLFCWUSKQB-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([S-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Loss on drying=<1 % (105°C)
Chloride (Cl)=<0.05 %
Calcium (Ca)=<50 ppm
Iron (Fe)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link