Sodium thiosulfate, 99%, for analysis, anhydrous, Thermo Scientific Chemicals
Sodium thiosulfate, 99%, for analysis, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium thiosulfate, 99%, for analysis, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 450622500
Price (USD)/ Each
23.65
Online exclusive
28.20 
Save 4.55 (16%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
23.65
Online exclusive
28.20 
Save 4.55 (16%)
Add to cart
Sodium thiosulfate, 99%, for analysis, anhydrous, Thermo Scientific Chemicals
Catalog number450622500
Price (USD)/ Each
23.65
Online exclusive
28.20 
Save 4.55 (16%)
-
Add to cart
Chemical Identifiers
IUPAC Namedisodium sulfanidesulfonate
Molecular FormulaNa2O3S2
InChI KeyAKHNMLFCWUSKQB-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([S-])(=O)=O
Molecular Weight (g/mol)158.10
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Loss on drying=<1 % (105°C)
Chloride (Cl)=<0.05 %
Calcium (Ca)=<50 ppm
Iron (Fe)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only