Thermo Scientific Chemicals

Calcium bromide hydrate, extra pure, Thermo Scientific Chemicals

Catalog number: 450632500
250 g, Each
Thermo Scientific Chemicals

Calcium bromide hydrate, extra pure, Thermo Scientific Chemicals

Catalog number: 450632500
250 g, Each
Quantity
Catalog number: 450632500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71626-99-8
IUPAC Name
calcium dibromide
Molecular Formula
Br2Ca
InChI Key
WGEFECGEFUFIQW-UHFFFAOYSA-L
SMILES
[Ca++].[Br-].[Br-]
Appearance (Form)
Crystalline powder and/or chunks
Solubility
(10 % in water) Clear colorless to slightly hazy
Appearance (Color)
White
Titration Complexometric
>=82 % (CaBr2)
Acidity/alkalinity
Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text