Thermo Scientific Chemicals

4-Hydrazinobenzoic acid hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 450670100
10 g, Each
Thermo Scientific Chemicals

4-Hydrazinobenzoic acid hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 450670100
10 g, Each
Quantity
Catalog number: 450670100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
24589-77-3
IUPAC Name
hydrogen 4-hydrazinylbenzoic acid chloride
Molecular Formula
C7H9ClN2O2
InChI Key
XHLQMKQBCHYRLC-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NNC1=CC=C(C=C1)C(O)=O
Appearance (Color)
Off-white to pink to red to brown
Appearance (Form)
Powder
Titration Argentometric
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text