Calcium carbonate, 99%, extra pure, Thermo Scientific Chemicals
Calcium carbonate, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium carbonate, 99%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
25 kg
Catalog number 450680050
Price (USD)/ Each
79.65
Online exclusive
88.00 
Save 8.35 (9%)
-
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)/ Each
79.65
Online exclusive
88.00 
Save 8.35 (9%)
Add to cart
Calcium carbonate, 99%, extra pure, Thermo Scientific Chemicals
Catalog number450680050
Price (USD)/ Each
79.65
Online exclusive
88.00 
Save 8.35 (9%)
-
Add to cart
Chemical Identifiers
CAS471-34-1
IUPAC Namecalcium carbonate
Molecular FormulaCCaO3
InChI KeyVTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES[Ca++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Complexometric98.5 to 101.5 %
Appearance (Color)White to almost white
Loss on drying=<2.0 % (1 g, 200°C)
Insoluble matter=<0.2 % (in dilute acetic acid)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only