Thermo Scientific Chemicals

D(+)-Glucose monohydrate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 450740250
25 kg, Fibre drum, Each
Thermo Scientific Chemicals

D(+)-Glucose monohydrate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 450740250
25 kg, Fibre drum, Each
Quantity
Packaging
Catalog number: 450740250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14431-43-7
IUPAC Name
(2S,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol hydrate
Molecular Formula
C6H14O7
InChI Key
OSNSWKAZFASRNG-WNFIKIDCSA-N
SMILES
O.OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O
Water
8 to 10 %
Appearance (Color)
White
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
HPLC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text