2-Amino-4-chlorobenzonitrile, 99%, Thermo Scientific Chemicals
2-Amino-4-chlorobenzonitrile, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-4-chlorobenzonitrile, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 450750100
Price (USD)/ Each
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS38487-86-4
IUPAC Name2-amino-4-chlorobenzonitrile
Molecular FormulaC7H5ClN2
InChI KeyUZHALXIAWJOLLR-UHFFFAOYSA-N
SMILESNC1=CC(Cl)=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow
Water=<0.5 %
Melting point157°C to 162°C
Infrared spectrumConforms
HPLC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only