Thermo Scientific Chemicals

2-Amino-4-chlorobenzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: 450750500
50 g, Each
Thermo Scientific Chemicals

2-Amino-4-chlorobenzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: 450750500
50 g, Each
Quantity
Catalog number: 450750500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38487-86-4
IUPAC Name
2-amino-4-chlorobenzonitrile
Molecular Formula
C7H5ClN2
InChI Key
UZHALXIAWJOLLR-UHFFFAOYSA-N
SMILES
NC1=CC(Cl)=CC=C1C#N
Appearance (Color)
Off-white to yellow
Water
=<0.5 %
Infrared spectrum
Conforms
HPLC
>=98.5 %
Melting point
157°C to 162°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text