3-Methylstyrene, 95%, Thermo Scientific Chemicals
3-Methylstyrene, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methylstyrene, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 450870010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS100-80-1
IUPAC Name1-ethenyl-3-methylbenzene
Molecular FormulaC9H10
InChI KeyJZHGRUMIRATHIU-UHFFFAOYSA-N
SMILESCC1=CC=CC(C=C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid
GC>=94.0 %
Infrared spectrumConforms
Stabilizer0.1 % 4-tert-Butylcatechol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only