Thermo Scientific Chemicals

3-Methylstyrene, 95%, Thermo Scientific Chemicals

Catalog number: 450870050
5 g, Each
Thermo Scientific Chemicals

3-Methylstyrene, 95%, Thermo Scientific Chemicals

Catalog number: 450870050
5 g, Each
Quantity
Catalog number: 450870050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
100-80-1
IUPAC Name
1-ethenyl-3-methylbenzene
Molecular Formula
C9H10
InChI Key
JZHGRUMIRATHIU-UHFFFAOYSA-N
SMILES
CC1=CC=CC(C=C)=C1
Appearance (Color)
Colorless to yellow
Appearance (Form)
Liquid
GC
>=94.0 %
Infrared spectrum
Conforms
Stabilizer
0.1 % 4-tert-Butylcatechol

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text