Thermo Scientific Chemicals

4-Iodophenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: 450900050
5 g, Each
Thermo Scientific Chemicals

4-Iodophenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: 450900050
5 g, Each
Quantity
Catalog number: 450900050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1798-06-7
IUPAC Name
2-(4-iodophenyl)acetic acid
Molecular Formula
C8H7IO2
InChI Key
FJSHTWVDFAUNCO-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=C(I)C=C1
Appearance (Form)
Powder or solid
Melting point
137°C to 143°C
Appearance (Color)
White to off-white to light brown
HPLC
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text