Sodium sulfate decahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Sodium sulfate decahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium sulfate decahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Packaging:
View all
Plastic Drum
Catalog number 451100050
Price (USD)/ Each
117.65
Online exclusive
131.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)/ Each
117.65
Online exclusive
131.00 
Save 13.35 (10%)
Add to cart
Sodium sulfate decahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Catalog number451100050
Price (USD)/ Each
117.65
Online exclusive
131.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7727-73-3
IUPAC Namedisodium decahydrate sulfate
Molecular FormulaH20Na2O14S
InChI KeyRSIJVJUOQBWMIM-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Titration after Ion exchange>=99.0 %
pH5.2 to 9.2 (5% soln. at 25°C)
Arsenic (As)=<0.5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only