Thermo Scientific Chemicals

Yeast extract

Catalog number: 451120010
1 kg, Each
Thermo Scientific Chemicals

Yeast extract

Catalog number: 451120010
1 kg, Each
Quantity
Catalog number: 451120010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
84604-16-0
Appearance (Color)
Yellow to brown
Appearance (Form)
Powder
Total nitrogen
>=9.0 % (on dry substance)
Amino nitrogen
>=3.0 % (on dry substance)
Loss on drying
=<6.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text