Thermo Scientific Chemicals

3-Aminooxetane, 95%, Thermo Scientific Chemicals

Catalog number: 451212500
250 mg, Each
Thermo Scientific Chemicals

3-Aminooxetane, 95%, Thermo Scientific Chemicals

Catalog number: 451212500
250 mg, Each
Quantity
Catalog number: 451212500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
21635-88-1
IUPAC Name
oxetan-3-amine
Molecular Formula
C3H7NO
InChI Key
OJEOJUQOECNDND-UHFFFAOYSA-N
SMILES
NC1COC1
Appearance (Form)
Liquid
GC
>=94.0 %
Appearance (Color)
Colorless to light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text