2-Amino-5-methoxybenzonitrile, 95%, Thermo Scientific Chemicals
2-Amino-5-methoxybenzonitrile, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-5-methoxybenzonitrile, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number 451340010
Price (USD)/ Each
95.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
112.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
95.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
112.00 
Save 16.35 (15%)
Add to cart
2-Amino-5-methoxybenzonitrile, 95%, Thermo Scientific Chemicals
Catalog number451340010
Price (USD)/ Each
95.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
112.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
CAS23842-82-2
IUPAC Name2-amino-5-methoxybenzonitrile
Molecular FormulaC8H8N2O
InChI KeySRWMPAZUWXLIPG-UHFFFAOYSA-N
SMILESCOC1=CC(C#N)=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point39°C to 45°C
Infrared spectrumConforms
GC>=94.0 %
Appearance (Color)White to yellow to brown
Appearance (Form)Powder or low melting solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only