2,4-Difluoro-3-nitrobenzonitrile, 97%, Thermo Scientific Chemicals
2,4-Difluoro-3-nitrobenzonitrile, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Difluoro-3-nitrobenzonitrile, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number 451420010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name2,4-difluoro-3-nitrobenzonitrile
Molecular FormulaC7H2F2N2O2
InChI KeyHESWWHRCQMLPFT-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=C(F)C=CC(C#N)=C1F
Molecular Weight (g/mol)184.10
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid or low melting solid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only