(1-Methyl-4-piperidinyl)methanamine, 97%, Thermo Scientific Chemicals
(1-Methyl-4-piperidinyl)methanamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(1-Methyl-4-piperidinyl)methanamine, 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
10 g
Catalog number 451460100
Price (USD)/ Each
789.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
789.00
Add to cart
(1-Methyl-4-piperidinyl)methanamine, 97%, Thermo Scientific Chemicals
Catalog number451460100
Price (USD)/ Each
789.00
-
Add to cart
Chemical Identifiers
IUPAC Name1-(1-methylpiperidin-4-yl)methanamine
Molecular FormulaC7H16N2
InChI KeyAGTPSAZJSOQXHJ-UHFFFAOYSA-N
SMILESCN1CCC(CN)CC1
Molecular Weight (g/mol)128.22
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Liquid
Appearance (Color)Colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only