5-Aminobenzothiazole, 95%, Thermo Scientific Chemicals
5-Aminobenzothiazole, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Aminobenzothiazole, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 451540050
Price (USD)/ Each
550.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
550.00
Add to cart
5-Aminobenzothiazole, 95%, Thermo Scientific Chemicals
Catalog number451540050
Price (USD)/ Each
550.00
-
Add to cart
Chemical Identifiers
CAS1123-93-9
IUPAC Name1,3-benzothiazol-5-amine
Molecular FormulaC7H6N2S
InChI KeyUJZYHMZRXGNDFB-UHFFFAOYSA-N
SMILESNC1=CC=C2SC=NC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown
Appearance (Form)Powder or solid
Infrared spectrumConforms
HPLC>=94.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only