Thermo Scientific Chemicals

L-Alanyl-L-glutamine, 97%

Catalog number: 451610010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

L-Alanyl-L-glutamine, 97%

Catalog number: 451610010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 451610010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39537-23-0
IUPAC Name
(2S)-2-[(2S)-2-aminopropanamido]-4-carbamoylbutanoic acid
Molecular Formula
C8H15N3O4
InChI Key
HJCMDXDYPOUFDY-WHFBIAKZSA-N
SMILES
C[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(O)=O
Appearance (Form)
Crystalline powder
Titration with HClO4
>=96.0 %
Heavy metals (as Pb)
=<10 ppm
Infrared spectrum
Conforms
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text