L-Alanyl-L-glutamine, 97%
L-Alanyl-L-glutamine, 97%
L-Alanyl-L-glutamine, 97%
L-Alanyl-L-glutamine, 97%
Thermo Scientific Chemicals

L-Alanyl-L-glutamine, 97%

CAS: 39537-23-0 | C8H15N3O4 | 217.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 451610050
Price (EUR)
127,50
Online Exclusive
150,00
Save 22,50 (15%)
Each
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
127,50
Online Exclusive
150,00
Save 22,50 (15%)
Each
Add to cart
Chemical Identifiers
CAS39537-23-0
IUPAC Name(2S)-2-[(2S)-2-aminopropanamido]-4-carbamoylbutanoic acid
Molecular FormulaC8H15N3O4
InChI KeyHJCMDXDYPOUFDY-WHFBIAKZSA-N
SMILESC[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=96.0 %
Heavy metals (as Pb)=<10 ppm
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Appearance (Color)White to off-white
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0464414Certificate of AnalysisJul 26, 2024451610050, 451610010
A0454867Certificate of AnalysisAug 25, 2023451610050, 451610010
A0420299Certificate of AnalysisJan 05, 2023451610050, 451610010
A0370236Certificate of AnalysisDec 14, 2022451610050, 451610010
A0414050Certificate of AnalysisDec 13, 2022451610050, 451610010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection