Thermo Scientific Chemicals

N-BOC-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 451720050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

N-BOC-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 451720050
5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 451720050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
286961-14-6
IUPAC Name
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine-1-carboxylate
Molecular Formula
C16H28BNO4
InChI Key
VVDCRJGWILREQH-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)N1CCC(=CC1)B1OC(C)(C)C(C)(C)O1
Appearance (Form)
Powder
Melting point
114°C to 118°C
Appearance (Color)
White to yellow to tan
Infrared spectrum
Conforms
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text