Chemical Identifiers
CAS286961-14-6
IUPAC Nametert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine-1-carboxylate
Molecular FormulaC16H28BNO4
InChI KeyVVDCRJGWILREQH-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCC(=CC1)B1OC(C)(C)C(C)(C)O1
View more