Thermo Scientific Chemicals

Ethanolamine, ACS reagent, Thermo Scientific Chemicals

Catalog number: 451760010
1 L, Each
Thermo Scientific Chemicals

Ethanolamine, ACS reagent, Thermo Scientific Chemicals

Catalog number: 451760010
1 L, Each
Quantity
Catalog number: 451760010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-43-5
IUPAC Name
2-aminoethan-1-ol
Molecular Formula
C2H7NO
InChI Key
HZAXFHJVJLSVMW-UHFFFAOYSA-N
SMILES
NCCO
Color scale
=<15 APHA
Water
=<0.30 %
Iron (Fe)
=<5 ppm
Appearance (Form)
Clear liquid
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text