Thermo Scientific Chemicals

Tetrabutylammonium fluoride, 75% solution in water, Thermo Scientific Chemicals

Catalog number: 451800250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetrabutylammonium fluoride, 75% solution in water, Thermo Scientific Chemicals

Catalog number: 451800250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 451800250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
429-41-4
IUPAC Name
tetrabutylazanium fluoride
Molecular Formula
C16H36FN
InChI Key
FPGGTKZVZWFYPV-UHFFFAOYSA-M
SMILES
[F-].CCCC[N+](CCCC)(CCCC)CCCC
Appearance (Color)
Colorless to light yellow
Titration with HClO4
70 to 80 %
Appearance (Form)
Viscous liquid
Water
20 to 30 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text