Thermo Scientific Chemicals

L-Alanine, 99%, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals

Catalog number: 451850025
2.5 kg, Each
Thermo Scientific Chemicals

L-Alanine, 99%, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals

Catalog number: 451850025
2.5 kg, Each
Quantity
Catalog number: 451850025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
56-41-7
IUPAC Name
(2S)-2-aminopropanoic acid
Molecular Formula
C3H7NO2
InChI Key
QNAYBMKLOCPYGJ-REOHCLBHSA-N
SMILES
C[C@H](N)C(O)=O
Appearance (Color)
Colorless to white to almost white
Infrared spectrum
Conforms
Appearance of solution
reference solution BY6
Specific optical rotation
+13.5° to +15.5° (20°C, 589 nm) (c=10, 6N HCl)
Appearance (Form)
Crystals or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text