Thermo Scientific Chemicals

Zinc acetate dihydrate, 97%, specified according to the requirements of USP, Thermo Scientific Chemicals

Catalog number: 451880025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Zinc acetate dihydrate, 97%, specified according to the requirements of USP, Thermo Scientific Chemicals

Catalog number: 451880025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 451880025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5970-45-6
IUPAC Name
zinc(2+) diacetate dihydrate
Molecular Formula
C4H10O6Zn
InChI Key
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
SMILES
O.O.[Zn++].CC([O-])=O.CC([O-])=O
Appearance (Form)
Crystalline powder or crystals
Titration Complexometric
98.0 to 102.0 %
pH
6.0 to 8.0 (5 % solution)
Chloride (Cl)
=<0.005 %
Sulfate (SO4)
=<0.01 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text