D(-)-Fructose, specified according the requirements of USP, Thermo Scientific Chemicals
D(-)-Fructose, specified according the requirements of USP, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D(-)-Fructose, specified according the requirements of USP, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Catalog number 451960010
Price (USD)/ Each
41.65
Online exclusive
48.90 
Save 7.25 (15%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
41.65
Online exclusive
48.90 
Save 7.25 (15%)
Add to cart
D(-)-Fructose, specified according the requirements of USP, Thermo Scientific Chemicals
Catalog number451960010
Price (USD)/ Each
41.65
Online exclusive
48.90 
Save 7.25 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
Molecular FormulaC6H12O6
InChI KeyBJHIKXHVCXFQLS-UYFOZJQFSA-N
SMILESOC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO
Molecular Weight (g/mol)180.16
View more
SpecificationsSpecification SheetSpecification Sheet
Assay98.0 to 102.0 %
Loss on drying=<0.5 %
AcidityPasses test
Arsenic (As)=<1 ppm
HydroxymethylfurfuralPasses test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only