Thermo Scientific Chemicals

D(-)-Fructose, specified according the requirements of USP, Thermo Scientific Chemicals

Catalog number: 451960050
5 kg, Each
Thermo Scientific Chemicals

D(-)-Fructose, specified according the requirements of USP, Thermo Scientific Chemicals

Catalog number: 451960050
5 kg, Each
Quantity
Catalog number: 451960050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57-48-7
IUPAC Name
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
Molecular Formula
C6H12O6
InChI Key
BJHIKXHVCXFQLS-UYFOZJQFSA-N
SMILES
OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO
Assay
98.0 to 102.0 %
Loss on drying
=<0.5 %
Acidity
Passes test
Arsenic (As)
=<1 ppm
Hydroxymethylfurfural
Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text