Melon™ Gel IgG Spin Purification Kit
Melon™ Gel IgG Spin Purification Kit
Thermo Scientific™

Melon™ Gel IgG Spin Purification Kit

The Thermo Scientific Melon Gel IgG Spin Purification Kit offers a simpler and faster alternative to traditional Protein A andRead more
Have Questions?
Catalog NumberQuantity
452063 mL
Catalog number 45206
Price (USD)
470.00
Each
-
Add to cart
Quantity:
3 mL
Price (USD)
470.00
Each
Add to cart
The Thermo Scientific Melon Gel IgG Spin Purification Kit offers a simpler and faster alternative to traditional Protein A and Protein G methods for purification of human, mouse and rabbit IgG from serum and other sources.

Features of Melon Gel IgG Purification Kits:

Simple, one-step protocol—IgG purification with Melon Gel Resin does not involve binding, washing, and multiple elution steps required by Protein A and Protein G
Rapid purification—antibody purification from serum is four to six times faster than Protein A or G methods
High recovery and purity—for many species of serum antibody recovery is greater than 90% with purity greater than 80%
Minimal dilution—reduced need to concentrate antibody before storage or use
Robust purification—Melon Gel IgG Purification Kits work with a wide range of antibodies including many that do not purify well on Protein A or Protein G
Gentle purification—No harsh antibody elution conditions helps retain IgG activity
Regenerable support—Melon Gel Resin can be used for multiple antibody purifications

Melon Gel IgG Purification Kits enable purification of antibodies from serum and other sample types. Melon Gel Resin binds and removes serum proteins, thereby allowing pure IgG to be collected in the flow-through fraction. The procedure is gentle because no harsh elution conditions are involved.

Melon Gel Resin contains a proprietary ligand that retains most protein found in serum, ascites and culture supernatants, while allowing IgG to pass through the support and be collected in the flow-through fraction. Because Melon Gel Resin is not a bind-and-release support, it is extremely fast and gentle to antibodies, resulting in antibody preparations of high purity and high activity.

Melon Gel Resin works with antibodies from a variety of species and subclasses, many of which do not purify efficiently with Protein A or Protein G. The resulting recovery and purity of the IgG isolated by this method rivals that obtained from the same samples using bind-and-release supports such as Protein A or Protein G. Furthermore, antibodies purified with Melon Gel Resin are recovered ready for use in immunoassays or subsequent conjugation or modification steps; there is no need to neutralize or desalt the recovered antibody to remove harsh elution buffers.

Melon Gel IgG Purification saves time compared to classical Protein A or Protein G methods without compromising the quality of the antibody. Compared to antibody affinity purification with Protein A or Protein G, Melon Gel IgG Purification is simpler, faster and provides greater yields and higher purity for a wider variety of antibodies species.

Related Products
Melon™ Gel IgG Purification Kit
Melon™ Gel Purification Buffer
Melon™ Gel Regenerant
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionMelon™ Gel IgG Spin Purification Kit, 3mL, Spin Column, Kit, Microcentrifuge Format, Affinity, Proprietary Resin Column Type, Proprietary Ligand Immobilized Phase, Sufficient For IgG Purification From Approximately 25 x 0.1mL Serum Samples
Purification TargetIgG Antibodies
Quantity3 mL
Column TypeProprietary Resin, Affinity
FormatKit, Microcentrifuge Spin Column
Product LineMelon™
Product TypeIgG Purification Kit
Stationary PhaseProprietary Ligand
Unit SizeEach
Contents & Storage
Sufficient For: IgG purification from approx. 25 x 0.1 mL serum samples (total approx. 25 mg IgG from serum)
• Melon Gel Support, 3 mL
• Melon Gel Purification Buffer, 100 mL
• Spin Columns, 0.8 mL, 27 columns
• Microcentrifuge Tubes, 2 mL, 30 tubes

Store at 4°C
Please enter sku to load default questions

Frequently asked questions (FAQs)

How can I determine when the Melon Gel Support is saturated?

For serum, a volume double the Melon Gel Support volume is the maximum amount that can be processed at one time. To determine if too much sample was processed, examine the purified IgG by SDS-PAGE for additional protein bands.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Can Melon Gel purify chicken IgY?

No. Thermo Scientific Thiophilic Adsorbent (Cat. Nos. 20500 and 44916) can be used to purify IgY from serum. The Chicken IgY Purification Kit (Cat. No. 44918) can be used to purify IgY from egg yolks.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Is the Melon Gel IgG Spin Purification Kit (Cat. No. 45206) a viable alternative to using traditional Protein A and Protein G methods for purification of IgG from serum?

Yes. With the Melon Gel IgG Spin Purification Kit, 90% recovery of IgG with greater than 80% purity are typically observed for a single purification. Although IgA and IgM may be present along with the purified IgG, a high percentage (> 80%) of IgA and IgM is retained on the gel support. This high binding of IgA and IgM to the gel support combined with the fact that IgA and IgM are present in low abundance in serum results in low levels of these antibody molecules in the purified IgG. Here are other advantages of using the Melon Gel IgG Spin Purification Kit over traditional Protein A or Protein G methods:
-Simple, one-step protocol - IgG purification with Melon Gel Resin does not involve binding, washing, and multiple elution steps required by Protein A and Protein G
-Rapid purification - antibody purification from serum is four to six times faster than Protein A or G methods
-Minimal dilution - reduced need to concentrate antibody before storage or use
-Robust purification - Melon Gel IgG Purification Kits work with a wide range of antibodies including many that do not purify well on Protein A or Protein G
-Gentle purification - No harsh antibody elution conditions helps retain IgG activity
-Regenerable support - Melon Gel Resin can be used for multiple antibody purifications

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How many times can I reuse the Melon Gel Support?

The Melon Gel Support can be used up to three times. Performing an ammonium sulfate precipitation on serum samples containing large amounts of non-antibody protein before purification improves purification efficiency and increases reusability.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

After Melon Gel purification, must I desalt or dialyze the purified antibody before using the Thermo Scientific Direct IP Kit (Cat. No. 26148)?

No. However, researchers may want to add an equal volume of binding buffer to elevate the pH towards neutral.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link