Thermo Scientific™

Pierce™ Saturated Ammonium Sulfate Solution

Catalog number: 45216
Have Questions?
Thermo Scientific™

Pierce™ Saturated Ammonium Sulfate Solution

Catalog number: 45216
Have Questions?
Catalog Number
45216
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
452161 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Saturated Ammonium Sulfate Solution is formulated and ready to use for simple immunoglobulin, antibody, or other protein fractionation applications based on selective precipitation, which is also called salting out.

Features of Saturated Ammonium Sulfate Solution:

Convenient—solution is ready to use; no preparation required
Concentrates—recover antibody in 100 mL from more than 1L of cell culture supernatant
Improves purity—partial-purification of antibodies by ammonium sulfate precipitation greatly improves the final purity that can be obtained with affinity purification
Extends column life—using pre-purified antibody samples instead of crude samples enables affinity columns to be used for many more cycles
Compatible—ammonium sulfate is easily removed by dialysis or desalting columns, eliminating any obstacles to downstream methods
Versatile—use for precipitating immunoglobulins or 'salting out' fractions of other sample types

The saturated ammonium sulfate solution (4.32M) enables removal of abundant proteins from serum, cell culture supernatant and ascites fluid. Slowly adding the solution to a protein sample results in decreased protein solubility and eventual protein precipitation as the charged ammonium sulfate ions compete with protein for water interaction. High molecular weight proteins, such as antibodies, are among the first to precipitate. When used in combination with antibody purification methods such as affinity chromatography, ammonium sulfate precipitation is ideal for achieving highly purified antibody samples.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Ammonium Sulfate Solution
Product Line
Pierce™
Column Type
Affinity
Reagent Type
Precipitation Reagent
Quantity
1 L
Form
Liquid

Contents & Storage

Upon receipt store at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text