Thermo Scientific Chemicals

1-BOC-3-(bromomethyl)azetidine, 95%, Thermo Scientific Chemicals

Catalog number: 452160010
1 g, Each
Thermo Scientific Chemicals

1-BOC-3-(bromomethyl)azetidine, 95%, Thermo Scientific Chemicals

Catalog number: 452160010
1 g, Each
Quantity
Catalog number: 452160010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
253176-93-1
IUPAC Name
tert-butyl 3-(bromomethyl)azetidine-1-carboxylate
Molecular Formula
C9H16BrNO2
InChI Key
PUKCUGDJEPVLPR-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)N1CC(CBr)C1
Appearance (Color)
White to yellow
Appearance (Form)
Low melting solid
Infrared spectrum
Conforms
GC
>=94.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text