Thermo Scientific Chemicals

Ethyl 3-(trifluoromethyl)pyrazole-4-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 452280250
25 g, Each
Thermo Scientific Chemicals

Ethyl 3-(trifluoromethyl)pyrazole-4-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 452280250
25 g, Each
Quantity
Catalog number: 452280250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
155377-19-8
Infrared spectrum
Conforms
Melting point
139°C to 145°C
Appearance (Color)
Off-white to yellow to tan
Appearance (Form)
Powder or solid
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text