Thermo Scientific Chemicals

2-Fluoropyridine-4-methanol, 97%, Thermo Scientific Chemicals

Catalog number: 452370050
5 g, Each
Thermo Scientific Chemicals

2-Fluoropyridine-4-methanol, 97%, Thermo Scientific Chemicals

Catalog number: 452370050
5 g, Each
Quantity
Catalog number: 452370050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
131747-60-9
IUPAC Name
(2-fluoropyridin-4-yl)methanol
Molecular Formula
C6H6FNO
InChI Key
IAFXOPAZVVMMLE-UHFFFAOYSA-N
SMILES
OCC1=CC(F)=NC=C1
Appearance (Form)
Powder
Appearance (Color)
White to off-white to pink
Infrared spectrum
Conforms
GC
>=96.0 %
Melting point
61°C to 67°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text