Thermo Scientific Chemicals

3-Fluoro-2-nitropyridine, 98%, Thermo Scientific Chemicals

Catalog number: 452450050
5 g, Each
Thermo Scientific Chemicals

3-Fluoro-2-nitropyridine, 98%, Thermo Scientific Chemicals

Catalog number: 452450050
5 g, Each
Quantity
Catalog number: 452450050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
54231-35-5
IUPAC Name
3-fluoro-2-nitropyridine
Molecular Formula
C5H3FN2O2
InChI Key
IJVFHCSUEBAAOZ-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=C(F)C=CC=N1
Appearance (Form)
Crystalline solid
NMR (1H-NMR)
Consistent with structure
Appearance (Color)
Yellow
Infrared spectrum
Conforms
Melting point
33°C to 37°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text