Thermo Scientific Chemicals

5-Amino-1-methyl-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog number: 452611000
100 g, Each
Thermo Scientific Chemicals

5-Amino-1-methyl-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog number: 452611000
100 g, Each
Quantity
Catalog number: 452611000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1192-21-8
IUPAC Name
1-methyl-1H-pyrazol-5-amine
Molecular Formula
C4H7N3
InChI Key
JESRNIJXVIFVOV-UHFFFAOYSA-N
SMILES
CN1N=CC=C1N
Appearance (Form)
Solid
Melting point
69°C to 77°C
Appearance (Color)
White to yellow
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text