Search Thermo Fisher Scientific
General Description
• Itraconazole is an aromatic ether that works as a strong inhibitor of P-glycoproteins and CYP3A4
• Itraconazole also acts as a hedgehog signaling inhibitor and P450 inhibitor
Applications
• Itraconazole can be exhibits antifungal activity against organisms including aspergillosis, blastomycosis, candidiasis, chromoblastomycosis, coccidioidomycosis, cryptococcosis, histoplasmosis, and sporotrichosis
• Itraconazole may be used to experimentally target the delivery of other molecules, such as plasmids, to cells
General References: