Thermo Scientific Chemicals

Trifluorothymidine, 98%, Thermo Scientific Chemicals

Catalog number: 452910010
1 g, Each
Thermo Scientific Chemicals

Trifluorothymidine, 98%, Thermo Scientific Chemicals

Catalog number: 452910010
1 g, Each
Quantity
Catalog number: 452910010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
70-00-8
IUPAC Name
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(trifluoromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Formula
C10H11F3N2O5
InChI Key
VSQQQLOSPVPRAZ-RRKCRQDMSA-N
SMILES
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C(=O)NC1=O)C(F)(F)F
Melting point
183°C to 189°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text