Chemical Identifiers
CAS70-00-8
IUPAC Name1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(trifluoromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC10H11F3N2O5
InChI KeyVSQQQLOSPVPRAZ-RRKCRQDMSA-N
SMILESOC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C(=O)NC1=O)C(F)(F)F
View more