(Isocyanoimino)triphenylphosphorane, 90%, Thermo Scientific Chemicals
(Isocyanoimino)triphenylphosphorane, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(Isocyanoimino)triphenylphosphorane, 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 452920050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
IUPAC Nameisocyano(triphenyl-λ⁵-phosphanylidene)amine
Molecular FormulaC19H15N2P
InChI KeyNIDTXBFHPXMXTR-UHFFFAOYSA-N
SMILES[C-]#[N+]N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)302.32
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=88 %
Appearance (Color)White to light yellow-orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only