4-(2'-Pyridylazo)resorcinol, 97+%, ACS reagent, Thermo Scientific Chemicals
4-(2'-Pyridylazo)resorcinol, 97+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(2'-Pyridylazo)resorcinol, 97+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number 452930010
Price (USD)/ Each
22.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
26.70 
Save 4.05 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
22.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
26.70 
Save 4.05 (15%)
Add to cart
4-(2'-Pyridylazo)resorcinol, 97+%, ACS reagent, Thermo Scientific Chemicals
Catalog number452930010
Price (USD)/ Each
22.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
26.70 
Save 4.05 (15%)
-
Add to cart
Chemical Identifiers
CAS1141-59-9
IUPAC Name(4Z)-3-hydroxy-4-[2-(pyridin-2-yl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Molecular FormulaC11H9N3O2
InChI KeyVLCAILLZPUINNF-LCYFTJDESA-N
SMILESOC1=CC(=O)C=C\C1=N\NC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO4>=97.0 %
Clarity of solutionPasses test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only