Thermo Scientific Chemicals

Dibutyl succinate, 99%, Thermo Scientific Chemicals

Catalog number: 453011000
100 mL, Each
Thermo Scientific Chemicals

Dibutyl succinate, 99%, Thermo Scientific Chemicals

Catalog number: 453011000
100 mL, Each
Quantity
Catalog number: 453011000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
141-03-7
IUPAC Name
1,4-dibutyl butanedioate
Molecular Formula
C12H22O4
InChI Key
YUXIBTJKHLUKBD-UHFFFAOYSA-N
SMILES
CCCCOC(=O)CCC(=O)OCCCC
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4280 to 1.4310 (20°C, 589 nm)
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text