6-Methylsalicylic acid, 98%, Thermo Scientific Chemicals
6-Methylsalicylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Methylsalicylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number 453060050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-hydroxy-6-methylbenzoic acid
Molecular FormulaC8H8O3
InChI KeyHCJMNOSIAGSZBM-UHFFFAOYSA-N
SMILESCC1=CC=CC(O)=C1C(O)=O
Molecular Weight (g/mol)152.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white to light brown
Appearance (Form)Powder
Infrared spectrumConforms
Melting point168°C to 176°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only