Thermo Scientific Chemicals

Graphene, Single Layer, Thermo Scientific Chemicals

Catalog number: 453150010
1 g, Each
Thermo Scientific Chemicals

Graphene, Single Layer, Thermo Scientific Chemicals

Catalog number: 453150010
1 g, Each
Quantity
Catalog number: 453150010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1034343-98-0
IUPAC Name
Graphene
Molecular Formula
Cx
SMILES
C
Synonym
methane collocarb carbene, 13c-labeled 13C-labeled carbene kosmos spheron magecol grosafe humenegro furnal
Oxygen (O)
7 to 7.5 %
Additional info
1-5 atomic layer graphene nanosheets
Additional info
lateral size: 0.5-5 µm
Surface area
650 to 750 m²/g
Conductivity
500 to 700 S/m

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text