Thermo Scientific Chemicals

Periodic acid, Thermo Scientific Chemicals

Catalog number: 453175000
500 g, Each
Thermo Scientific Chemicals

Periodic acid, Thermo Scientific Chemicals

Catalog number: 453175000
500 g, Each
Quantity
Catalog number: 453175000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10450-60-9
IUPAC Name
tetrahydroxyiodous acid
Molecular Formula
H5IO6
InChI Key
TWLXDPFBEPBAQB-UHFFFAOYSA-N
SMILES
O[I](O)(O)(O)(O)=O
Sodium (Na)
=<100 ppm
Zinc (Zn)
=<50 ppm
Nitrate (NO3)
=<100 ppm
Sulfate (SO4)
=<100 ppm
Chloride and Chlorate (as Cl)
=<200 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text