Potassium nitrite, 97%, extra pure, Thermo Scientific Chemicals
Potassium nitrite, 97%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium nitrite, 97%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number 453190050
Price (USD)/ Each
334.65
Online exclusive
372.00 
Save 37.35 (10%)
-
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)/ Each
334.65
Online exclusive
372.00 
Save 37.35 (10%)
Add to cart
Potassium nitrite, 97%, extra pure, Thermo Scientific Chemicals
Catalog number453190050
Price (USD)/ Each
334.65
Online exclusive
372.00 
Save 37.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7758-09-0
IUPAC Namepotassium nitrite
Molecular FormulaKNO2
InChI KeyBXNHTSHTPBPRFX-UHFFFAOYSA-M
SMILES[K+].[O-]N=O
View more
SpecificationsSpecification SheetSpecification Sheet
Arsenic (As)=<1 ppm
Heavy metals (as Pb)=<10 ppm
Titration excess KMnO496.0 to 104.0 %
Appearance (Form)Powder or crystals
Insoluble matter=<0.01 % (10 % in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only