Thermo Scientific Chemicals

3-Maleimidopropionic acid N-hydroxysuccinimide ester, 99%, Thermo Scientific Chemicals

Catalog number: 453250010
1 g, Each
Thermo Scientific Chemicals

3-Maleimidopropionic acid N-hydroxysuccinimide ester, 99%, Thermo Scientific Chemicals

Catalog number: 453250010
1 g, Each
Quantity
Catalog number: 453250010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
55750-62-4
IUPAC Name
2,5-dioxopyrrolidin-1-yl 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoate
Molecular Formula
C11H10N2O6
InChI Key
JKHVDAUOODACDU-UHFFFAOYSA-N
SMILES
O=C(CCN1C(=O)C=CC1=O)ON1C(=O)CCC1=O
Appearance (Color)
White to light yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text