Thermo Scientific Chemicals

3-Methoxypropionic acid, 97+%, Thermo Scientific Chemicals

Catalog number: 453320500
50 mL, Each
Thermo Scientific Chemicals

3-Methoxypropionic acid, 97+%, Thermo Scientific Chemicals

Catalog number: 453320500
50 mL, Each
Quantity
Catalog number: 453320500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2544-06-1
IUPAC Name
3-methoxypropanoic acid
Molecular Formula
C4H8O3
InChI Key
YSIKHBWUBSFBRZ-UHFFFAOYSA-N
SMILES
COCCC(O)=O
Appearance (Color)
Colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text