Imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals
Imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 453360010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1423031-35-9
IUPAC Nameimidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride
Molecular FormulaC8H7ClN2O2
InChI KeyOQGRMPZJJDBYBL-UHFFFAOYSA-N
SMILESCl.OC(=O)C1=CC2=NC=CN2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
Titration Argentometric>=96.0 to =<104.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only