Thermo Scientific Chemicals

1-Tetrahydropyran-4-yl-1H-pyrazol-4-ylamine, 97%, Thermo Scientific Chemicals

Catalog number: 453570050
5 g, Each
Thermo Scientific Chemicals

1-Tetrahydropyran-4-yl-1H-pyrazol-4-ylamine, 97%, Thermo Scientific Chemicals

Catalog number: 453570050
5 g, Each
Quantity
Catalog number: 453570050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1190380-49-4
IUPAC Name
1-(oxan-4-yl)-1H-pyrazol-4-amine
Molecular Formula
C8H13N3O
InChI Key
GZZNBQLBGSVOOZ-UHFFFAOYSA-N
SMILES
NC1=CN(N=C1)C1CCOCC1
Appearance (Color)
White to light yellow to pink
GC
>=96.0 %
Infrared spectrum
Conforms
NMR (1H-NMR)
Consistent with structure
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text