Thermo Scientific Chemicals

4-Methyl-1,3-thiazole-5-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 453640010
1 g, Each
Thermo Scientific Chemicals

4-Methyl-1,3-thiazole-5-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 453640010
1 g, Each
Quantity
Catalog number: 453640010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
953070-51-4
IUPAC Name
4-methyl-1,3-thiazole-5-sulfonyl chloride
Molecular Formula
C4H4ClNO2S2
InChI Key
AWTJUYXVSWZCIK-UHFFFAOYSA-N
SMILES
CC1=C(SC=N1)S(Cl)(=O)=O
Appearance (Form)
Low melting solid
Infrared spectrum
Conforms
GC
>=96.0 %
Appearance (Color)
White to yellow to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text